missing translation for 'onlineSavingsMsg'
Learn More

3-Methoxyphenyl isothiocyanate, 98%

Product Code. p-7022359
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
This item is not returnable. View return policy

Product Code. 11437397

Brand: Thermo Scientific Alfa Aesar A15090.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 3125-64-2 | C8H7NOS | 165.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3125-64-2
Molecular Formula C8H7NOS
Molecular Weight (g/mol) 165.21
MDL Number MFCD00040335
InChI Key WHBYCPUKGYEYFU-UHFFFAOYSA-N
Synonym 3-methoxyphenyl isothiocyanate, 3-methoxyphenylisothiocyanate, 1-isothiocyanato-3-methoxy-benzene, 3-isothiocyanatoanisole, acmc-1cldj, m-methoxyphenyl isothiocyanate, 3-methoxy phenyl isothiocyanate, 3-methoxy-phenyl isothiocyanate, #
PubChem CID 137832
IUPAC Name 1-isothiocyanato-3-methoxybenzene
SMILES COC1=CC=CC(=C1)N=C=S

Specifications

Density 1.179
Boiling Point 133°C to 134°C (10 mmHg)
Flash Point >110°C (230°F)
Odor Pungent
Refractive Index 1.639
Quantity 5 g
UN Number UN2922
Beilstein 775987
Sensitivity Moisture sensitive
Formula Weight 165.22
Percent Purity 98%
Chemical Name or Material 3-Methoxyphenyl isothiocyanate
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.