missing translation for 'onlineSavingsMsg'
Learn More

3-(Methoxymethylene)-2(3H)-benzofuranone, TRC

Product Code. 30470665
Change view
Click to view available options
Quantity:
100 mg
1 g
5 g
Unit Size:
100mg
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30470665 100 mg 100mg
30472289 1 g 1g
30475588 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30470665 Supplier Toronto Research Chemicals Supplier No. TRCM264230100MG
 Restricted Item
Estimated Shipment: 07-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 40800-90-6
Molecular Formula C10H8O3
Molecular Weight (g/mol) 176.17
IUPAC Name (3Z)-3-(methoxymethylidene)-1-benzofuran-2-one
SMILES CO\C=C\1/C(=O)Oc2ccccc12

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C10H8O3/c1-12-6-8-7-4-2-3-5-9(7)13-10(8)11/h2-6H,1H3/b8-6-
Formula Weight 176.0473
Chemical Name or Material 3-(Methoxymethylene)-2(3H)-benzofuranone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.