missing translation for 'onlineSavingsMsg'
Learn More

3'-Methoxybiphenyl-4-sulfonyl chloride, 96%

Product Code. p-7031936
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15426017 250 mg 250mg
15406017 1 g 1g
15416017 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15426017 Supplier Thermo Scientific Alfa Aesar Supplier No. H33211.MD
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 186550-26-5 | C13H11ClO3S | 282.738 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 186550-26-5
Molecular Formula C13H11ClO3S
Molecular Weight (g/mol) 282.738
MDL Number MFCD06409270
InChI Key DVQBUJTYQXKLKZ-UHFFFAOYSA-N
Synonym 3'-methoxy-biphenyl-4-sulfonyl chloride, 3'-methoxybiphenyl-4-sulfonyl chloride, 3'-methoxy 1,1'-biphenyl-4-sulfonyl chloride, 3'-methoxy-1,1'-biphenyl-4-sulfonyl chloride, 1,1'-biphenyl-4-sulfonylchloride, 3'-methoxy, 4-3-methoxyphenyl benzenesulfonyl chloride, 3'-methoxy-biphenyl-4-sulfonylchloride, 3/'-methoxybiphenyl-4-sulfonyl chloride
PubChem CID 10826798
IUPAC Name 4-(3-methoxyphenyl)benzenesulfonyl chloride
SMILES COC1=CC=CC(=C1)C2=CC=C(C=C2)S(=O)(=O)Cl

Specifications

Melting Point 80°C to 84°C
Quantity 250 mg
UN Number UN3261
Sensitivity Moisture sensitive
Formula Weight 282.74
Percent Purity 96%
Chemical Name or Material 3'-Methoxybiphenyl-4-sulfonyl chloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.