missing translation for 'onlineSavingsMsg'
Learn More

3-Methoxybenzyl chloride, 98+%

Product Code. 11480463
Change view
Click to view available options
Quantity:
5 g
25 g
50 g
250 g
Unit Size:
250g
25g
50g
5g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11480463 250 g 250g
11379469 5 g 5g
11389469 25 g 25g
11470463 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 11480463 Supplier Thermo Scientific Alfa Aesar Supplier No. A12630.30
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 824-98-6 | C8H9ClO | 156.61 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 824-98-6
Molecular Formula C8H9ClO
Molecular Weight (g/mol) 156.61
MDL Number MFCD00000907
InChI Key VGISFWWEOGVMED-UHFFFAOYSA-N
Synonym 3-methoxybenzyl chloride, 1-chloromethyl-3-methoxybenzene, m-methoxybenzyl chloride, m-chloromethyl anisole, benzene, 1-chloromethyl-3-methoxy, anisole, m-chloromethyl, 3-methoxybenzylchloride, 3-chloromethyl anisole, 3-methoxy benzyl chloride, benzyl chloride,3-methoxy
PubChem CID 69994
IUPAC Name 1-(chloromethyl)-3-methoxybenzene
SMILES COC1=CC=CC(CCl)=C1

Specifications

Density 1.078
Boiling Point 122°C to 123°C (12 mmHg)
Flash Point 101°C (213°F)
Refractive Index 1.544
Quantity 250 g
UN Number UN1760
Beilstein 636684
Sensitivity Moisture sensitive
Formula Weight 156.61
Percent Purity ≥98%
Chemical Name or Material 3-Methoxybenzyl chloride
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.