missing translation for 'onlineSavingsMsg'
Learn More

3-Methoxybenzyl chloride, 97%

Product Code. 10164020 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10164020 100 g 100g
10338480 5 g 5g
10062520 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10164020 Supplier Thermo Scientific Acros Supplier No. 197861000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 824-98-6 | C8H9ClO | 156.61 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 824-98-6
Molecular Formula C8H9ClO
Molecular Weight (g/mol) 156.61
MDL Number MFCD00000907
InChI Key VGISFWWEOGVMED-UHFFFAOYSA-N
Synonym 3-methoxybenzyl chloride, 1-chloromethyl-3-methoxybenzene, m-methoxybenzyl chloride, m-chloromethyl anisole, benzene, 1-chloromethyl-3-methoxy, anisole, m-chloromethyl, 3-methoxybenzylchloride, 3-chloromethyl anisole, 3-methoxy benzyl chloride, benzyl chloride,3-methoxy
PubChem CID 69994
IUPAC Name 1-(chloromethyl)-3-methoxybenzene
SMILES COC1=CC=CC(CCl)=C1

Specifications

CAS Min % 96.0
CAS Max % 100.0
Density 1.0700g/mL
Boiling Point 124°C (13.0 mmHg)
Flash Point 101°C
Assay Percent Range 96% min. (GC)
Packaging Glass Bottle
Linear Formula CH3OC6H4CH2Cl
Refractive Index 1.5430 to 1.545
Quantity 100 g
Beilstein 06, I, 189
Specific Gravity 1.07
Formula Weight 156.61
Percent Purity 97%
Chemical Name or Material 3-Methoxybenzyl chloride, 97%
Show More Show Less
compliance-icons
Product Identifier
  • 3-Methoxybenzyl chloride
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.