missing translation for 'onlineSavingsMsg'
Learn More

3-Methoxybenzoyl chloride, 99%

Product Code. 11495597
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11495597 250 g 250g
11475597 10 g 10g
11485597 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11495597 Supplier Thermo Scientific Alfa Aesar Supplier No. B23571.30
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 1711-05-3 | C8H7ClO2 | 170.592 g/mol

3-Methoxybenzoyl chloride is used in the synthesis of 2-arylbenzofuran-based molecules that act against Alzheimer’s disease, combating symptoms. In addition, it aids in the optimization of dihydropyrrolopyrimidine inhibitors against PI3K (phosphoinositide 3-kinase) resulting in tumor growth inhibition.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Soluble in chloroform. Reacts with water.

Notes
Moisture Sensitive. Store under dry inert gas. Store away from moisture, strong bases and oxidizing agents. Protect from humidity and water.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1711-05-3
Molecular Formula C8H7ClO2
Molecular Weight (g/mol) 170.592
MDL Number MFCD00000673
InChI Key RUQIUASLAXJZIE-UHFFFAOYSA-N
Synonym m-anisoyl chloride, benzoyl chloride, 3-methoxy, m-methoxybenzoyl chloride, 3-anisoyl chloride, m-anisic acid chloride, 3-methoxy-benzoyl chloride, benzoyl chloride, m-methoxy, 3-methoxybenzoic acid chloride, m-anisoylchloride, 3-anisoylchlorid
PubChem CID 74374
IUPAC Name 3-methoxybenzoyl chloride
SMILES COC1=CC=CC(=C1)C(=O)Cl

Specifications

Density 1.214
Boiling Point 123°C to 125°C (15 mmHg)
Flash Point 92°C (197°F)
Refractive Index 1.5586
Quantity 250 g
UN Number UN1729
Beilstein 386659
Sensitivity Moisture sensitive
Solubility Information Soluble in chloroform. Reacts with water.
Formula Weight 170.6
Percent Purity 99%
Chemical Name or Material 3-Methoxybenzoyl chloride
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.