missing translation for 'onlineSavingsMsg'
Learn More

3-Methoxybenzonitrile, 98%

Product Code. 11461197
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11461197 5 g 5g
11471197 25 g 25g
11481197 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11461197 Supplier Thermo Scientific Alfa Aesar Supplier No. B24171.06
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 1527-89-5 | C8H7NO | 133.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1527-89-5
Molecular Formula C8H7NO
Molecular Weight (g/mol) 133.15
MDL Number MFCD00001801
InChI Key KLXSUMLEPNAZFK-UHFFFAOYSA-N
Synonym m-cyanoanisole, benzonitrile, 3-methoxy, m-anisonitrile, m-methoxybenzonitrile, 3-cyanoanisole, 3-anisonitrile, benzonitrile, m-methoxy, 3-methoxy-benzonitrile, 3-methoxy benzonitrile, m-methoxybenzontrile
PubChem CID 73712
IUPAC Name 3-methoxybenzonitrile
SMILES COC1=CC=CC(=C1)C#N

Specifications

Melting Point 22°C to 24°C
Boiling Point 110°C to 111°C (12 mmHg)
Flash Point 105°C (221°F)
Odor Almond-like
Quantity 5 g
UN Number UN3439
Beilstein 1932666
Formula Weight 133.15
Percent Purity 98%
Chemical Name or Material 3-Methoxybenzonitrile

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.