missing translation for 'onlineSavingsMsg'
Learn More

3-Methoxybenzoic acid, 99%

Product Code. 11409672
Change view
Click to view available options
Quantity:
50 g
250 g
1000 g
Unit Size:
1000g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11409672 250 g 250g
11499662 50 g 50g
11489662 1000 g 1000g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11409672 Supplier Thermo Scientific Alfa Aesar Supplier No. A10767.30
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 586-38-9 | C8H8O3 | 152.149 g/mol

3-Methoxybenzoic acid is used in the synthesis and characterization of 3-methoxybenzoates of europium (III) and gadolinium (III)1. It was used in conversion of aromatic carboxylic acids into methyl esters and reduction to the corresponding primary alcohols using a sodium borohydride-THF-methanol system

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Methoxybenzoic acid is used in the synthesis and characterization of 3-methoxybenzoates of europium (III) and gadolinium (III)1. It was used in conversion of aromatic carboxylic acids into methyl esters and reduction to the corresponding primary alcohols using a sodium borohydride-THF-methanol system

Solubility
Soluble in 95% ethanol (50 mg/ml), water (2 mg/ml at 25°C) and methanol.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with bases and oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 586-38-9
Molecular Formula C8H8O3
Molecular Weight (g/mol) 152.149
MDL Number MFCD00002499
InChI Key XHQZJYCNDZAGLW-UHFFFAOYSA-N
Synonym m-anisic acid, m-methoxybenzoic acid, benzoic acid, 3-methoxy, 3-anisic acid, meta-anisic acid, 3-methoxy-benzoic acid, unii-e2i36fh6qz, m-methylsalicylic acid, e2i36fh6qz, chembl22425
PubChem CID 11461
IUPAC Name 3-methoxybenzoic acid
SMILES COC1=CC=CC(=C1)C(=O)O

Specifications

Melting Point 104°C to 108°C
Boiling Point 170°C to 172°C (10 mmHg)
Odor Odorless
Quantity 250 g
Beilstein 508838
Solubility Information Soluble in 95% ethanol (50mg/ml),water (2mg/ml at 25°C) and methanol.
Formula Weight 152.15
Percent Purity 99%
Chemical Name or Material 3-Methoxybenzoic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.