missing translation for 'onlineSavingsMsg'
Learn More

3-Methoxybenzhydrazide, 98+%

Product Code. 11450017
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11450017 5 g 5g
11460017 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11450017 Supplier Thermo Scientific Alfa Aesar Supplier No. B20956.06
 Restricted Item
Estimated Shipment: 20-07-2026
Add to basket
This item is not returnable. View return policy

CAS: 5785-06-8 | C8H10N2O2 | 166.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5785-06-8
MDL Number MFCD00007601

Specifications

Melting Point 90°C to 94°C
Molecular Formula C10H16
Quantity 5 g
Beilstein 2092135
Synonym 1s-2,2-dimethyl-3-methylidenebicyclo 2.2.1 heptane, s-camphene, 1s,4r-2,2-dimethyl-3-methylene-bicyclo 2.2.1 heptane
InChI Key CRPUJAZIXJMDBK-UHFFFAOYNA-N
SMILES CC1(C2CCC(C2)C1=C)C
IUPAC Name (1S,4S)-3,3-dimethyl-2-methylidenebicyclo[2.2.1]heptane
Molecular Weight (g/mol) 136.24
PubChem CID 6560154
Formula Weight 166.18
Percent Purity ≥98%
Chemical Name or Material 3-Methoxybenzhydrazide
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.