missing translation for 'onlineSavingsMsg'
Learn More

3-Methoxy Desloratadine, TRC

Product Code. 30477318
Change view
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30477318 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30477318 Supplier Toronto Research Chemicals Supplier No. TRCM2624205MG
 Restricted Item
Estimated Shipment: 19-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

CAS 165739-63-9
Molecular Formula C20 H21 Cl N2 O
Quantity 5 mg
InChI Formula InChI=1S/C20H21ClN2O/c1-24-17-11-15-3-2-14-10-16(21)4-5-18(14)19(20(15)23-12-17)13-6-8-22-9-7-13/h4-5,10-12,22H,2-3,6-9H2,1H3
SMILES COc1cnc2C(=C3CCNCC3)c4ccc(Cl)cc4CCc2c1
IUPAC Name 8-chloro-3-methoxy-11-piperidin-4-ylidene-5,6-dihydrobenzo[1,2]cyclohepta[2,4-b]pyridine
Molecular Weight (g/mol) 340.85
Formula Weight 340.1342
Chemical Name or Material 3-Methoxy Desloratadine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.