missing translation for 'onlineSavingsMsg'
Learn More

3-Methoxy-5-(trifluoromethyl)phenylacetonitrile, 97%, Thermo Scientific™

Product Code. 15497845
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15497845 1 g 1g
15407855 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15497845 Supplier Thermo Scientific Alfa Aesar Supplier No. H31621.03

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 916421-02-8
Molecular Formula C10H8F3NO
Molecular Weight (g/mol) 215.18
MDL Number MFCD09025415
InChI Key IZJPZDDRQKCSEL-UHFFFAOYSA-N
Synonym 3-methoxy-5-trifluoromethyl phenylacetonitrile, 2-3-methoxy-5-trifluoromethyl phenyl acetonitrile, 3-methoxy-5-trifluoromethyl phenyl acetonitrile, benzeneacetonitrile, 3-methoxy-5-trifluoromethyl, 2-5-methoxy-3-trifluoromethyl phenyl ethanenitrile
PubChem CID 46737580
IUPAC Name 2-[3-methoxy-5-(trifluoromethyl)phenyl]acetonitrile
SMILES COC1=CC(CC#N)=CC(=C1)C(F)(F)F

Specifikationer

Refractive Index 1.475
Quantity 1 g
UN Number UN3276
Formula Weight 215.18
Percent Purity 97%
Chemical Name or Material 3-Methoxy-5-(trifluoromethyl)phenylacetonitrile
Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.