missing translation for 'onlineSavingsMsg'
Learn More

3-Methoxy-5-(trifluoromethyl)phenol, 97%, Thermo Scientific™

Product Code. 15405617
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15405617 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15405617 Supplier Thermo Scientific Alfa Aesar Supplier No. H32848.MD

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 349-56-4
Molecular Formula C8H7F3O2
Molecular Weight (g/mol) 192.137
MDL Number MFCD09025418
InChI Key DFOFNISQFZKBEY-UHFFFAOYSA-N
Synonym 3-methoxy-5-trifluoromethyl-phenol, 3-methoxy-5-trifluoromethyl phenol, 3-hydroxy-5-methoxybenzotrifluoride, 3-hydroxy-5-trifluoromethyl anisole, 5-methoxy-3-trifluoromethyl phenol, 3-hydroxy-5-methoxy-benzotrifluoride, 3-methoxy-5-hydroxybenzotrifluoride, 3-hydroxy-5-methoxy-1-trifluoromethylbenzene
PubChem CID 22571708
IUPAC Name 3-methoxy-5-(trifluoromethyl)phenol
SMILES COC1=CC(=CC(=C1)O)C(F)(F)F

Specifications

Melting Point 74°C to 76°C
Quantity 250 mg
Formula Weight 192.14
Percent Purity 97%
Chemical Name or Material 3-Methoxy-5-(trifluoromethyl)phenol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.