missing translation for 'onlineSavingsMsg'
Learn More

3-Methoxy-4-(trifluoromethyl)benzyl bromide, 97%, Thermo Scientific™

Product Code. 15476767
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
Product Code. Quantity unitSize
15476767 1 g 1g
1 options
This item is not returnable. View return policy

Product Code. 15476767

Brand: Thermo Scientific Alfa Aesar H33880.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 853367-87-0
Molecular Formula C9H8BrF3O
Molecular Weight (g/mol) 269.061
MDL Number MFCD18398596
InChI Key MUVUCILNTIWNQL-UHFFFAOYSA-N
Synonym 3-methoxy-4-trifluoromethyl benzyl bromide, 4-bromomethyl-2-methoxy-1-trifluoromethyl benzene
PubChem CID 66523605
IUPAC Name 4-(bromomethyl)-2-methoxy-1-(trifluoromethyl)benzene
SMILES COC1=C(C=CC(=C1)CBr)C(F)(F)F

Specifications

Quantity 1 g
UN Number UN3261
Formula Weight 269.06
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 3-Methoxy-4-(trifluoromethyl)benzyl bromide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.