missing translation for 'onlineSavingsMsg'
Learn More

3-Methoxy-4-(methoxycarbonyl)phenylboronic acid, 97%, Thermo Scientific™

Product Code. 11906151 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11906151 1 g 1g
11916151 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11906151 Supplier Thermo Scientific Acros Supplier No. 445480010

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 603122-41-4
Molecular Formula C9H11BO5
Molecular Weight (g/mol) 210
InChI Key YCXPWNGIPLGCOJ-UHFFFAOYSA-N
Synonym 3-methoxy-4-methoxycarbonylphenylboronic acid, 3-methoxy-4-methoxycarbonyl phenylboronic acid, 4-methoxycarbonyl-3-methoxyphenylboronic acid, 3-methoxy-4-methoxycarbonyl phenyl boronic acid, 3-methoxy-4-methoxycarbonyl-phenyl boronic acid, 3-methoxy-4-methoxycarbonyl benzeneboronic acid, 3-methoxy-4-methoxycarbonylphenylboronicacid, ycxpwngiplgcoj-uhfffaoysa-n, methyl 4-borono-2-methoxybenzoate
PubChem CID 17750256

Specifications

Assay Percent Range 96% min. (HPLC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 210
Percent Purity 97%
Chemical Name or Material 3-Methoxy-4-(methoxycarbonyl)phenylboronic acid
compliance-icons
Product Identifier
  • 3-Methoxy-4-(methoxycarbonyl)phenylboronic acid
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.