missing translation for 'onlineSavingsMsg'
Learn More

3-Methoxy-3-methylbutyl Acetate, TRC

Product Code. 30468512
Change view
Click to view available options
Quantity:
250 mg
500 mg
2.5 g
Unit Size:
2.5g
250mg
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30468512 500 mg 500mg
30470388 2.5 g 2.5g
30466502 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30468512 Supplier Toronto Research Chemicals Supplier No. TRCM331833500MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 103429-90-9
Molecular Formula C8H16O3
Molecular Weight (g/mol) 160.21
IUPAC Name (3-methoxy-3-methylbutyl) acetate
SMILES COC(C)(C)CCOC(=O)C

Specifications

Quantity 500 mg
InChI Formula InChI=1S/C8H16O3/c1-7(9)11-6-5-8(2,3)10-4/h5-6H2,1-4H3
Formula Weight 160.1099
Chemical Name or Material 3-Methoxy-3-methylbutyl Acetate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.