missing translation for 'onlineSavingsMsg'
Learn More

3-Methoxy-2-(trifluoromethyl)benzaldehyde, 97%, Thermo Scientific™

Product Code. 15438275
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1214384-89-0
Molecular Formula C9H7F3O2
Molecular Weight (g/mol) 204.148
MDL Number MFCD13185677
InChI Key HSPNVFXNJYKWSK-UHFFFAOYSA-N
Synonym 3-methoxy-2-trifluoromethyl benzaldehyde
PubChem CID 73012639
IUPAC Name 3-methoxy-2-(trifluoromethyl)benzaldehyde
SMILES COC1=CC=CC(=C1C(F)(F)F)C=O

Specifications

Color Yellow
Refractive Index 1.489
Quantity 250 mg
Sensitivity Air sensitive
Formula Weight 204.15
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 3-Methoxy-2-(trifluoromethyl)benzaldehyde
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'