missing translation for 'onlineSavingsMsg'
Learn More

3-Methoxy-2-methyl-N-phenylbenzamide-d3, TRC

Product Code. 30468447
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Quantity:
100 mg
missing translation for 'unitSize':
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity missing translation for 'unitSize'
30468447 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 missing translation for 'options'
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30468447 missing translation for 'mfr' Toronto Research Chemicals missing translation for 'supplierNo' TRCM264602100MG
 Restricted Item
Estimated Shipment: 11-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 3-Methoxy-2-methyl-d3-N-phenylbenzamide
CAS 1020719-60-1
Molecular Formula C15 2H3 H12 N O2
Quantity 100 mg
InChI Formula InChI=1S/C15H15NO2/c1-11-13(9-6-10-14(11)18-2)15(17)16-12-7-4-3-5-8-12/h3-10H,1-2H3,(H,16,17)/i1D3
SMILES [2H]C([2H])([2H])c1c(OC)cccc1C(=O)Nc2ccccc2
IUPAC Name 3-methoxy-N-phenyl-2-(trideuteriomethyl)benzamide
Molecular Weight (g/mol) 244.3
Formula Weight 244.1291
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.