missing translation for 'onlineSavingsMsg'
Learn More

3-Methoxy-1,2-propanediol, TRC

Product Code. 17928314
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17928314 1 g 1g
17989064 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17928314 Supplier Toronto Research Chemicals Supplier No. TRCM2665401G
 Restricted Item
Estimated Shipment: 24-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 623-39-2
Molecular Formula C4 H10 O3
Molecular Weight (g/mol) 106.12
Synonym 1-Methoxy-2,3-propanediol, 3-Methoxy-1,2-dihydroxypropane, Glycerin-α-monomethyl ether, Glycerol 1-monomethyl ether, NSC 6752
IUPAC Name 3-methoxypropane-1,2-diol
SMILES COCC(O)CO

Specifications

Quantity 1 g
InChI Formula InChI=1S/C4H10O3/c1-7-3-4(6)2-5/h4-6H,2-3H2,1H3
Formula Weight 106.063
Chemical Name or Material 3-Methoxy-1,2-propanediol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.