missing translation for 'onlineSavingsMsg'
Learn More

3-(Methacryloyloxy)propyltris(trimethylsiloxy)silane, 98%

Product Code. 11389325
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11389325 100 g 100g
11369325 5 g 5g
11379325 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11389325 Supplier Thermo Scientific Alfa Aesar Supplier No. L16682.22
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 17096-07-0 | C16H38O5Si4 | 422.815 g/mol

3-(Methacryloyloxy)propyltris(trimethylsiloxy) silane acts as a tetramethylsilane-protected silicate compound utilized for proteomics research. It is also used as a silane coupling agent, silane adhesion promoter, silane hydrophobing agent, silane dispersing agent and silane crosslinking agent. It is also employed as a silane moisture scavenger, polypropylene catalyst, silicate stabilizer, polyurethane endcapper silane, silane drying agent, silane curing agent and silane reinforcer. Further, it serves as a silylating agent, silane reducing agent, silyl building block and synthons in synthetic chemistry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-(Methacryloyloxy)propyltris(trimethylsiloxy) silane acts as a tetramethylsilane-protected silicate compound utilized for proteomics research. It is also used as a silane coupling agent, silane adhesion promoter, silane hydrophobing agent, silane dispersing agent and silane crosslinking agent. It is also employed as a silane moisture scavenger, polypropylene catalyst, silicate stabilizer, polyurethane endcapper silane, silane drying agent, silane curing agent and silane reinforcer. Further, it serves as a silylating agent, silane reducing agent, silyl building block and synthons in synthetic chemistry.

Solubility
Immiscible with water.

Notes
Incompatible with strong oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 17096-07-0
Molecular Formula C16H38O5Si4
Molecular Weight (g/mol) 422.815
MDL Number MFCD00053871
InChI Key BESKSSIEODQWBP-UHFFFAOYSA-N
Synonym 3-methacryloyloxy propyltris trimethylsiloxy silane, 3-tris trimethylsiloxy silyl propyl methacrylate, 3-1,1,1,5,5,5-hexamethyl-3-trimethylsilyl oxy trisiloxan-3-yl propyl methacrylate, unii-d32s78j7t8, 3-methacryloyloxypropyl tris trimethylsiloxy silane, 3-methacryloxypropyltris trimethylsiloxy silane, 3-tris trimethylsilyloxy silyl propyl methacrylate, methacryloxypropyltris trimethylsiloxy silane, 3-3,3,3-trimethyl-1,1-bis trimethylsilyl oxy disiloxanyl propyl methacrylate
PubChem CID 123371
IUPAC Name 3-tris(trimethylsilyloxy)silylpropyl 2-methylprop-2-enoate
SMILES CC(=C)C(=O)OCCC[Si](O[Si](C)(C)C)(O[Si](C)(C)C)O[Si](C)(C)C

Specifications

Boiling Point 112°C to 115°C (0.2mmHg)
Quantity 100 g
Flash Point 201°C (393°F)
Solubility Information Immiscible with water.
Formula Weight 422.82
Percent Purity 98%
Refractive Index 1.4175
Density 0.93
Chemical Name or Material 3-(Methacryloyloxy)propyltris(trimethylsiloxy)silane
compliance-icons
Product Identifier
  • 3-(Methacryloyloxy)propyltris(trimethylsiloxy)silane
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contain: 3-(Methacryloyloxy)propyltris(trimethylsiloxy)silane

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.