Learn More
3-(Methacryloyloxy)propyltrimethoxysilane, 97%, Thermo Scientific Chemicals
£23.00 - £111.00
Chemical Identifiers
CAS | 2530-85-0 |
---|---|
Molecular Formula | C10H20O5Si |
Molecular Weight (g/mol) | 248.35 |
MDL Number | MFCD00008593 |
InChI Key | XDLMVUHYZWKMMD-UHFFFAOYSA-N |
Synonym | 3-trimethoxysilyl propyl methacrylate, 3-methacryloxypropyltrimethoxysilane, methacryloxypropyltrimethoxysilane, dynasylan memo, silicone a-174, union carbide a-174, mops-m, silane a-174, nuca 174, 2-propenoic acid, 2-methyl-, 3-trimethoxysilyl propyl ester |
PubChem CID | 17318 |
IUPAC Name | 3-trimethoxysilylpropyl 2-methylprop-2-enoate |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15493157
|
Thermo Scientific Alfa Aesar
A17714.18 |
50 g |
£23.00
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11385119
|
Thermo Scientific Alfa Aesar
A17714.22 |
100 g |
£33.90
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15403167
|
Thermo Scientific Alfa Aesar
A17714.30 |
250 g | |||||||
11395119
|
Thermo Scientific Alfa Aesar
A17714.36 |
500 g |
£111.00
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
3-(Methacryloyloxy)propyltrimethoxysilane (MPS) acts as a functional comonomer, which is used to prepare polystyrene latex having silanol by emulsion polymerization. It is used in the preparation of polymers in association with other monomers like vinyl acetate, acrylic acid and methyl acrylic acid used in coating, adhesive and sealing agents, which provides excellent adhesion and durability. The composite materials made from unsaturated polyester by using MPS improve mechanical property.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications3-(Methacryloyloxy)propyltrimethoxysilane (MPS) acts as a functional comonomer, which is used to prepare polystyrene latex having silanol by emulsion polymerization. It is used in the preparation of polymers in association with other monomers like vinyl acetate, acrylic acid and methyl acrylic acid used in coating, adhesive and sealing agents, which provides excellent adhesion and durability. The composite materials made from unsaturated polyester by using MPS improve mechanical property.
Solubility
Miscible with water.
Notes
Moisture sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents, strong acids and strong bases.
Chemical Identifiers
2530-85-0 | |
248.35 | |
XDLMVUHYZWKMMD-UHFFFAOYSA-N | |
17318 |
C10H20O5Si | |
MFCD00008593 | |
3-trimethoxysilyl propyl methacrylate, 3-methacryloxypropyltrimethoxysilane, methacryloxypropyltrimethoxysilane, dynasylan memo, silicone a-174, union carbide a-174, mops-m, silane a-174, nuca 174, 2-propenoic acid, 2-methyl-, 3-trimethoxysilyl propyl ester | |
3-trimethoxysilylpropyl 2-methylprop-2-enoate |
Specifications
190°C | |
C10H20O5Si | |
1952435 | |
3-trimethoxysilyl propyl methacrylate, 3-methacryloxypropyltrimethoxysilane, methacryloxypropyltrimethoxysilane, dynasylan memo, silicone a-174, union carbide a-174, mops-m, silane a-174, nuca 174, 2-propenoic acid, 2-methyl-, 3-trimethoxysilyl propyl ester | |
XDLMVUHYZWKMMD-UHFFFAOYSA-N | |
3-trimethoxysilylpropyl 2-methylprop-2-enoate | |
17318 | |
97% | |
Moisture sensitive | |
3-(Methacryloyloxy)propyltrimethoxysilane |
2530-85-0 | |
MFCD00008593 | |
92°C | |
Miscible with water. | |
CO[Si](CCCOC(=O)C(C)=C)(OC)OC | |
248.35 | |
248.35 | |
1.431 | |
1.045 |
Safety and Handling
GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'einecsNumber' : 219-785-8
missing translation for 'rtecsNumber' : UC0230000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only