missing translation for 'onlineSavingsMsg'
Learn More

(3-Mercaptopropyl)trimethoxysilane, 95%

Product Code. 11490148
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11490148 100 g 100g
11480148 25 g 25g
11400158 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11490148 Supplier Thermo Scientific Alfa Aesar Supplier No. B23726.22
  • ONLINE EXCLUSIVE PRICE
    £61.35
    £49.65 / 100g
    Save £11.70
    19% Off
View all promotions
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 4420-74-0 | C6H16O3SSi | 196.336 g/mol

(3-Mercaptopropyl)trimethoxysilane is used as a capping molecule for cadmium sulphide nanocrystals. It is used in the preparation of multi-walled carbon nanotubes and bilayer modified gold electrode. It acts as a linker molecule used in the preparation of gold multilayers and as a molecular adhesive in the fabrication of vapor-deposited gold electrodes on glass substrates. It is also used as a precursor for silica.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(3-Mercaptopropyl)trimethoxysilane is used as a capping molecule for cadmium sulphide nanocrystals. It is used in the preparation of multi-walled carbon nanotubes and bilayer modified gold electrode. It acts as a linker molecule used in the preparation of gold multilayers and as a molecular adhesive in the fabrication of vapor-deposited gold electrodes on glass substrates. It is also used as a precursor for silica.

Solubility
Miscible with methanol, ethanol, isopropanol, acetone, benzene, toluene and xylene.

Notes
Air and moisture sensitive. Incompatible with strong oxidizing agents, strong acids, alcohols and water. Decomposes in water.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4420-74-0
Molecular Formula C6H16O3SSi
Molecular Weight (g/mol) 196.336
MDL Number MFCD00004901
InChI Key UUEWCQRISZBELL-UHFFFAOYSA-N
Synonym trimethoxysilylpropanethiol, 3-mercaptopropyltrimethoxysilane, 3-mercaptopropyl trimethoxysilane, 1-propanethiol, 3-trimethoxysilyl, silquest a 189, 3-trimethoxysilyl propanethiol, prosil 196, silane a 189, union carbide a-189, sila-ace s 810
PubChem CID 20473
IUPAC Name 3-trimethoxysilylpropane-1-thiol
SMILES CO[Si](CCCS)(OC)OC

Specifications

Boiling Point 213°C to 215°C
Melting Point <-50°C
Quantity 100 g
Flash Point 96°C (204°F)
Refractive Index 1.443
Sensitivity Air and moisture sensitive
Density 1.05
Linear Formula (CH3O)3Si(CH2)3SH
UN Number UN3082
Beilstein 2038119
Solubility Information Miscible with methanol,ethanol,isopropanol,acetone,benzene,toluene and xylene.
Formula Weight 196.34
Percent Purity 95%
Chemical Name or Material (3-Mercaptopropyl)trimethoxysilane
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • (3-Mercaptopropyl)trimethoxysilane
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Skin sensitiser Category 1
Hazard Statement
  • H302-Harmful if swallowed.
  • H317-May cause an allergic skin reaction.
  • H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: 3-Mercaptopropyltrimethoxysilane

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.