missing translation for 'onlineSavingsMsg'
Learn More

(3-Mercaptopropyl)trimethoxysilane, 85%, Tech.

Product Code. 10219273 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
50 g
250 g
Packaging:
Glass bottle
Unit Size:
250g
50g
This item is not returnable. View return policy

Product Code. 10219273

Brand: Thermo Scientific Acros 174652500

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 4420-74-0 | C6H16O3SSi | 196.34 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4420-74-0
Molecular Formula C6H16O3SSi
Molecular Weight (g/mol) 196.34
MDL Number MFCD00004901
InChI Key UUEWCQRISZBELL-UHFFFAOYSA-N
Synonym trimethoxysilylpropanethiol, 3-mercaptopropyltrimethoxysilane, 3-mercaptopropyl trimethoxysilane, 1-propanethiol, 3-trimethoxysilyl, silquest a 189, 3-trimethoxysilyl propanethiol, prosil 196, silane a 189, union carbide a-189, sila-ace s 810
PubChem CID 20473
IUPAC Name 3-trimethoxysilylpropane-1-thiol
SMILES CO[Si](CCCS)(OC)OC

Specifications

CAS Min % 80.0
CAS Max % 100.0
Color Colorless to Yellow
Density 1.0500g/mL
Boiling Point 215°C
Flash Point 88°C
Infrared Spectrum Authentic
Assay Percent Range 80% min. (GC)
Packaging Glass bottle
Linear Formula HSCH2CH2CH2Si(OCH3)3
Refractive Index 1.4410 to 1.443
Quantity 250 g
Beilstein 10, 148A
Specific Gravity 1.05
Solubility Information Solubility in water: decomposes. Other solubilities: soluble in methanol,ethanol,isopropanol,,mineral spirits,acetone,benzene,toluene and,xylene
Viscosity 2 mPa.s ( 20°C)
Formula Weight 196.34
Percent Purity 85%
Grade Technical
Physical Form Liquid
Chemical Name or Material (3-Mercaptopropyl)trimethoxysilane
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • (3-Mercaptopropyl)trimethoxysilane
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Respiratory sensitiser Category 1
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H411-Toxic to aquatic life with long lasting effects.
  • H312-Harmful in contact with skin.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.