missing translation for 'onlineSavingsMsg'
Learn More

(3-Mercaptopropyl)methyldimethoxysilane, 95%

Codice prodotto. 11344598
Cambia vista
Click to view available options
Quantity:
10 mL
50 mL
250 mL
1 L
Dimensione della confezione:
1 L
10mL
250mL
50mL
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Quantity unitSize
11344598 10 mL 10mL
11324598 50 mL 50mL
11354598 250 mL 250mL
11334598 1 L 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
Codice prodotto. 11344598 Fornitore Thermo Scientific Alfa Aesar N. del fornitore 042426.AB
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Visualizzare prodotti alternativi

Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso

CAS: 31001-77-1 | C6H16O2SSi | 180.337 g/mol

(3-Mercaptopropyl)methyldimethoxysilane is used as coupling agent to improve the adhesion of sulfur cured elastomers (polysulfide, polyurethane sealants) to inorganic fillers (such as silica, glass, clay, mica), fiberglass and surfaces. It is also used as a pretreatment on minerals or added at the time of compounding and to improve low-rolling resistance in silica-reinforced tire tread compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Identificatori chimici

CAS 31001-77-1
Molecular Formula C6H16O2SSi
Molecular Weight (g/mol) 180.337
MDL Number MFCD00053923
InChI Key IKYAJDOSWUATPI-UHFFFAOYSA-N
Synonym 3-mercaptopropylmethyldimethoxysilane, 3-mercaptopropylmethyldimethoxy silane, 3-dimethoxymethylsilyl-1-propanethiol, 3-mercaptopropyl methyldimethoxysilane, dimethoxy-3-mercaptopropyl methylsilane, 3-dimethoxy methyl silyl propane-1-thiol, acmc-1aiak, ksc496q0j, 3-methyldimethoxysilyl propylmercaptan
PubChem CID 2759524
IUPAC Name 3-[dimethoxy(methyl)silyl]propane-1-thiol
SMILES CO[Si](C)(CCCS)OC

Specifica

Boiling Point 96°C (30mmHg)
Color Colorless
Physical Form Liquid
Quantity 10 mL
Linear Formula HSCH2CH2CH2Si(OCH3)2CH3
Flash Point 93°C (199°F)
Solubility Information Insoluble in water.
Formula Weight 180.34
Percent Purity 95%
Refractive Index 1.45
Sensitivity Moisture sensitive
Density 1.000 g/mL
Chemical Name or Material (3-Mercaptopropyl)methyldimethoxysilane
Vedi altri risultati Mostra meno risultati

RUO – Research Use Only

Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.