missing translation for 'onlineSavingsMsg'
Learn More

(3-Mercaptopropyl)methyldimethoxysilane, 95%

Product Code. 11354598
Change view
Click to view available options
Quantity:
10 mL
50 mL
250 mL
1 L
Unit Size:
1 L
10mL
250mL
50mL
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11354598 250 mL 250mL
11344598 10 mL 10mL
11324598 50 mL 50mL
11334598 1 L 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 11354598 Supplier Thermo Scientific Alfa Aesar Supplier No. 042426.AK
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 31001-77-1 | C6H16O2SSi | 180.337 g/mol

(3-Mercaptopropyl)methyldimethoxysilane is used as coupling agent to improve the adhesion of sulfur cured elastomers (polysulfide, polyurethane sealants) to inorganic fillers (such as silica, glass, clay, mica), fiberglass and surfaces. It is also used as a pretreatment on minerals or added at the time of compounding and to improve low-rolling resistance in silica-reinforced tire tread compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 31001-77-1
Molecular Formula C6H16O2SSi
Molecular Weight (g/mol) 180.337
MDL Number MFCD00053923
InChI Key IKYAJDOSWUATPI-UHFFFAOYSA-N
Synonym 3-mercaptopropylmethyldimethoxysilane, 3-mercaptopropylmethyldimethoxy silane, 3-dimethoxymethylsilyl-1-propanethiol, 3-mercaptopropyl methyldimethoxysilane, dimethoxy-3-mercaptopropyl methylsilane, 3-dimethoxy methyl silyl propane-1-thiol, acmc-1aiak, ksc496q0j, 3-methyldimethoxysilyl propylmercaptan
PubChem CID 2759524
IUPAC Name 3-[dimethoxy(methyl)silyl]propane-1-thiol
SMILES CO[Si](C)(CCCS)OC

Specifications

Boiling Point 96°C (30mmHg)
Color Colorless
Physical Form Liquid
Quantity 250 mL
Linear Formula HSCH2CH2CH2Si(OCH3)2CH3
Flash Point 93°C (199°F)
Solubility Information Insoluble in water.
Formula Weight 180.34
Percent Purity 95%
Refractive Index 1.45
Sensitivity Moisture sensitive
Density 1.000 g/mL
Chemical Name or Material (3-Mercaptopropyl)methyldimethoxysilane
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.