missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ 3-Mercaptopicolinic acid hydrochloride

Product Code. 18717786
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18717786 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18717786 Supplier Tocris Bioscience™ Supplier No. 7577/50

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Phospoenolpyruvate carboxykinase (PEPCK) inhibitor; inhibits gluconeogenesis

3-Mercaptopicolinic acid hydrochloride is a phospoenolpyruvate carboxykinase (PEPCK) inhibitor (IC50 = 7.5 μM at hPEPCK). Inhibits glucose synthesis. In vitro, 3-Mercaptopicolinic acid reduces proliferation of Treg cells, induces myogenic differentiation of C2C12 cells, and reduces colony growth formation of MCF7 cells. Also sensitizes melanoma cells to Vemurafenib (Cat. No. 7309) via inhibition of cytoplasmic PEPCK.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 3-Mercapto-2-pyridinecarboxylic acid hydrochloride
CAS 320386-54-7
Quantity 50 mg
Target PCK1
Molecular Formula C6H5NO2S.HCl
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.