missing translation for 'onlineSavingsMsg'
Learn More

3-Mercaptopicolinic Acid Hydrochloride, TRC

Product Code. 30475764
Change view
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30475764 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30475764 Supplier Toronto Research Chemicals Supplier No. TRCM257000500MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifikationer

Chemical Name or Material 3-Mercaptopicolinic Acid, Hydrochloride
CAS 320386-54-7
Molecular Formula C6 H5 N O2 S . Cl H
Quantity 500 mg
Synonym 3-Mercapto-2-pyridinecarboxylic acid hydrochloride
InChI Formula InChI=1S/C6H5NO2S.ClH/c8-6(9)5-4(10)2-1-3-7-5;/h1-3,10H,(H,8,9);1H
SMILES Cl.OC(=O)c1ncccc1S
IUPAC Name 3-sulfanylpyridine-2-carboxylic acid;hydrochloride
Molecular Weight (g/mol) 191.64
Formula Weight 190.9808
Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.