missing translation for 'onlineSavingsMsg'
Learn More

3-Mercapto-3-methyl-1-butanol, TRC

Product Code. 30474547
Change view
Click to view available options
Quantity:
25 mg
50 mg
250 mg
Unit Size:
250mg
25mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30474547 50 mg 50mg
30467966 250 mg 250mg
30467040 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30474547 Supplier Toronto Research Chemicals Supplier No. TRCM25758350MG
 Restricted Item
Estimated Shipment: 19-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 34300-94-2
Molecular Formula C5H12OS
Molecular Weight (g/mol) 120.21
Synonym 1-Butanol, 3-mercapto-3-methyl- (8CI, 9CI, ACI), 3-Mercapto-3-methyl-1-butanol (ACI), 3-Mercapto-3-methylbutanol, 3-Methyl-3-sulfanylbutan-1-ol, 3-Sulfanyl-3-methylbutan-1-ol
IUPAC Name 3-methyl-3-sulfanylbutan-1-ol
SMILES CC(C)(S)CCO

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C5H12OS/c1-5(2,7)3-4-6/h6-7H,3-4H2,1-2H3
Formula Weight 120.0609
Chemical Name or Material 3-Mercapto-3-methyl-1-butanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.