missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Mercapto-1-propanol, TRC
Click to view available options
Quantity:
1 g
Unit Size:
1g
Specifications
Specifications
| Chemical Name or Material | 3-Mercapto-1-propanol |
| CAS | 19721-22-3 |
| Molecular Formula | C3H8OS |
| Quantity | 1 g |
| Synonym | 3-Mercaptopropanol |
| InChI Formula | InChI=1S/C3H8OS/c4-2-1-3-5/h4-5H,1-3H2 |
| SMILES | OCCCS |
| IUPAC Name | 3-sulfanylpropan-1-ol |
| Molecular Weight (g/mol) | 92.16 |
| Formula Weight | 92.0296 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction