missing translation for 'onlineSavingsMsg'
Learn More

3-Mercapto-1-propanol, 97+%

Product Code. 10045253 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10045253 5 g Glass bottle 5g
10184713 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10045253 Supplier Thermo Scientific Acros Supplier No. 388570050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 19721-22-3 | C3H8OS | 92.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 19721-22-3
Molecular Formula C3H8OS
Molecular Weight (g/mol) 92.16
MDL Number MFCD00192262
InChI Key SHLSSLVZXJBVHE-UHFFFAOYSA-N
Synonym 3-mercapto-1-propanol, 1-propanol, 3-mercapto, 3-mercaptopropanol, 3-hydroxypropanethiol, 3-hydroxypropyl mercaptan, 3-mecapto-1-propanol, 3-mecaptopropan-1-ol, 1-mercapto-3-propanol, 3-hydroxypropylmercaptan, 3-mercaptopropan-1-ol
PubChem CID 88211
IUPAC Name 3-sulfanylpropan-1-ol
SMILES C(CO)CS

Specifications

CAS Min % 97.0
CAS Max % 100.0
Color Colorless
Density 1.0620g/mL
Boiling Point 106°C (50.0 mmHg)
Flash Point 85°C
Infrared Spectrum Authentic
Assay Percent Range 97% min. (GC)
Packaging Glass bottle
Linear Formula SH(CH2)3OH
Refractive Index 1.4950 to 1.497
Quantity 5 g
Specific Gravity 1.062
Solubility Information Solubility in water: slightly soluble
Formula Weight 92.16
Percent Purity 97+%
Physical Form Liquid
Chemical Name or Material 3-Mercapto-1-propanol
Show More Show Less
compliance-icons
Product Identifier
  • 3-Mercapto-1-propanol
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H311-Toxic in contact with skin.
  • H335-May cause respiratory irritation.
  • H301-Toxic if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.