missing translation for 'onlineSavingsMsg'
Learn More

3-Mercapto-1-hexanol, 96%

Product Code. 10051881 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 mL
25 mL
Packaging:
Glass bottle
Unit Size:
25mL
5mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10051881 25 mL Glass bottle 25mL
10410252 5 mL Glass bottle 5mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10051881 Supplier Thermo Scientific Acros Supplier No. 347560250
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 51755-83-0 | C6H14OS | 134.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 51755-83-0
Molecular Formula C6H14OS
Molecular Weight (g/mol) 134.23
MDL Number MFCD00792515
InChI Key TYZFMFVWHZKYSE-UHFFFAOYSA-N
Synonym 3-mercapto-1-hexanol, 3-mercaptohexanol, 3-mercaptohexan-1-ol, 1-hexanol, 3-mercapto, 3-thiohexan-1-ol, 3-sulphanylhexan-1-ol, 3-sulfanyl-1-hexanol, 3-thiohexanol, acmc-209kvm, 3-mercapto-hexan-1-ol
PubChem CID 521348
ChEBI CHEBI:77690
IUPAC Name 3-sulfanylhexan-1-ol
SMILES CCCC(CCO)S

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color Colorless
Density 0.9700g/mL
Boiling Point 250°C
Flash Point >100°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula H3C(CH2)2CH(SH)(CH2)2OH
Refractive Index 1.4790 to 1.481
Quantity 25 mL
Specific Gravity 0.97
Solubility Information Solubility in water: insoluble in water.,soluble in organic solvents (most)
Formula Weight 134.23
Percent Purity 96%
Physical Form Liquid
Chemical Name or Material 3-Mercapto-1-hexanol
Show More Show Less
compliance-icons
Product Identifier
  • 3-Mercapto-1-hexanol
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.