missing translation for 'onlineSavingsMsg'
Learn More

3-Mercapto-1-hexanol, 96%

Product Code. 11455077
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11455077 25 g 25g
11445077 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11455077 Supplier Thermo Scientific Alfa Aesar Supplier No. B21965.14
 Restricted Item
Estimated Shipment: 28-09-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 51755-83-0 | C6H14OS | 134.237 g/mol

3-mercapto-1-hexanol (3MH) are compounds potentially responsible for the development of blackcurrant aroma in red wine. 3-mercapto-1-hexanol was the most important odorant of Grenache rosé wine was with a deep impact on the wine fruity and citric notes. 3-Mercaptohexanol is an thiol compound found in various grape juices.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-mercapto-1-hexanol (3MH) are compounds potentially responsible for the development of blackcurrant aroma in red wine. 3-mercapto-1-hexanol was the most important odorant of Grenache rosé wine was with a deep impact on the wine fruity and citric notes. 3-Mercaptohexanol is an thiol compound found in various grape juices.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 51755-83-0
Molecular Formula C6H14OS
Molecular Weight (g/mol) 134.237
MDL Number MFCD00792515
InChI Key TYZFMFVWHZKYSE-UHFFFAOYSA-N
Synonym 3-mercapto-1-hexanol, 3-mercaptohexanol, 3-mercaptohexan-1-ol, 1-hexanol, 3-mercapto, 3-thiohexan-1-ol, 3-sulphanylhexan-1-ol, 3-sulfanyl-1-hexanol, 3-thiohexanol, acmc-209kvm, 3-mercapto-hexan-1-ol
PubChem CID 521348
ChEBI CHEBI:77690
IUPAC Name 3-sulfanylhexan-1-ol
SMILES CCCC(CCO)S

Specifications

Density 0.974
Boiling Point 250°C
Flash Point >100°C (212°F)
Refractive Index 1.48
Quantity 25 g
Beilstein 1846873
Solubility Information Insoluble in water.
Formula Weight 134.24
Percent Purity 96%
Chemical Name or Material 3-Mercapto-1-hexanol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.