missing translation for 'onlineSavingsMsg'
Learn More

3-Mercapto-1,2-propanediol, 90%

Product Code. p-7024385
Click to view available options
Quantity:
50 g
250 g
Unit Size:
250g
50g
This item is not returnable. View return policy

Product Code. 11315249

Brand: Thermo Scientific Alfa Aesar A18948.18

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 96-27-5 | C3H8O2S | 108.16 g/mol

It generally used as a size-regulating capping agent for nanocrystals. 1-Thioglycerol is also used in bacteriology experiments to induce the synthesis of porphyrin compounds in aerobically growing Escherichia coli. Also used to study the pH-sensitive photoluminescence of aqueous thiol-capped CdTe nanocrystals. 1-Thioglycerol is used to develop and test thiol functionalized copolymers. 1-Thioglycerol is used as a post-modification agent in the generation of non-standard peptide foldamers.Potential substitute for 2-mercaptoethanol, probe for the study of lymphocyte activation.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It generally used as a size-regulating capping agent for nanocrystals. 1-Thioglycerol is also used in bacteriology experiments to induce the synthesis of porphyrin compounds in aerobically growing Escherichia coli. Also used to study the pH-sensitive photoluminescence of aqueous thiol-capped CdTe nanocrystals. 1-Thioglycerol is used to develop and test thiol functionalized copolymers. 1-Thioglycerol is used as a post-modification agent in the generation of non-standard peptide foldamers.Potential substitute for 2-mercaptoethanol, probe for the study of lymphocyte activation.

Solubility
Slightly soluble in water and benzene. Soluble in acetone. Insoluble in ether.

Notes
Air Sensitive and Hygroscopic, store away from water/moisture and air. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store under dry inert gas.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 96-27-5
Molecular Formula C3H8O2S
Molecular Weight (g/mol) 108.16
MDL Number MFCD00004879
InChI Key PJUIMOJAAPLTRJ-UHFFFAOYNA-N
Synonym 3-mercapto-1,2-propanediol, monothioglycerol, 1-thioglycerol, thioglycerol, thioglycerine, thioglycerin, thiovanol, 3-mercaptopropane-1,2-diol, alpha-thioglycerol, glycerol-1-thiol
PubChem CID 7291
ChEBI CHEBI:74537
IUPAC Name 3-sulfanylpropane-1,2-diol
SMILES OCC(O)CS

Specifications

Density 1.247
Boiling Point 117°C to 119°C (5 mmHg)
Flash Point 149°C (300°F)
Linear Formula HOCH2CH(OH)CH2SH
Refractive Index 1.511
Quantity 50 g
Beilstein 1732046
Sensitivity Air sensitive; Hygroscopic
Merck Index 14,9335
Solubility Information Slightly soluble in water and benzene. Soluble in acetone. Insoluble in ether.
Formula Weight 108.16
Percent Purity ≥90%
Chemical Name or Material 3-Mercapto-1,2-propanediol
Show More Show Less
compliance-icons
Product Identifier
  • N-Ethylmaleimide
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Skin sensitiser Category 1
  • Specific target organ toxicity Category 3
Hazard Statement
  • H302-Harmful if swallowed.
  • H311-Toxic in contact with skin.
  • H315-Causes skin irritation.
  • H317-May cause an allergic skin reaction.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : Maleic acid N-ethylimide

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.