missing translation for 'onlineSavingsMsg'
Learn More

3-Mercapto-1,2,4-triazole, 98%

Product Code. 11424474
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11424474 100 g 100g
11404474 5 g 5g
11414474 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11424474 Supplier Thermo Scientific Alfa Aesar Supplier No. A13120.22
 Restricted Item
Estimated Shipment: 08-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 3179-31-5 | C2H3N3S | 101.13 g/mol

3-Mercapto-1,2,4-triazole was used in a study to design a surface enhanced Raman scattering based probe for fast and accurate detection of DNA markers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Mercapto-1,2,4-triazole was used in a study to design a surface enhanced Raman scattering based probe for fast and accurate detection of DNA markers.

Solubility
Soluble in hot water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3179-31-5
Molecular Formula C2H3N3S
Molecular Weight (g/mol) 101.13
MDL Number MFCD00005229
InChI Key AFBBKYQYNPNMAT-UHFFFAOYSA-N
Synonym 1h-1,2,4-triazole-3-thiol, 3-mercapto-1,2,4-triazole, 1,2,4-triazole-3-thiol, mercaptotriazole, 4h-1,2,4-triazole-3-thiol, s-triazole-3-thiol, 3-mercapto-1h-1,2,4-triazole, 1,2,4-triazole-5-thiol, 1,3,4-triazine-2-thiol, 1,3,4-triazole-2-thiol
PubChem CID 2723802
SMILES S=C1NNC=N1

Specifications

Melting Point 217°C to 224°C
Quantity 100 g
Beilstein 107731
Solubility Information Soluble in hot water.
IUPAC Name 2,3-dihydro-1H-1,2,4-triazole-3-thione
Formula Weight 101.13
Percent Purity 98%
Chemical Name or Material 3-Mercapto-1,2,4-triazole

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.