missing translation for 'onlineSavingsMsg'
Learn More

3-Maleimidopropionic Acid, TRC

Product Code. 30483005
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30483005 5 g 5g
30477836 250 mg 250mg
30470586 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30483005 Supplier Toronto Research Chemicals Supplier No. TRCM1407505G
 Restricted Item
Estimated Shipment: 21-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7423-55-4
Molecular Formula C7 H7 N O4
Molecular Weight (g/mol) 169.13
IUPAC Name 3-(2,5-dioxopyrrol-1-yl)propanoic acid
SMILES OC(=O)CCN1C(=O)C=CC1=O

Specifications

Quantity 5 g
InChI Formula InChI=1S/C7H7NO4/c9-5-1-2-6(10)8(5)4-3-7(11)12/h1-2H,3-4H2,(H,11,12)
Formula Weight 169.0375
Chemical Name or Material 3-Maleimidopropionic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.