missing translation for 'onlineSavingsMsg'
Learn More

3-Maleimidopropionic acid, 95%, Thermo Scientific Chemicals

Product Code. 15428098
Click to view available options
Quantity:
1 g
5 g

3-Maleimidopropionic acid is a sulfhydryl reactive heterobifunctional crosslinking reagent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Maleimidopropionic acid is a sulfhydryl reactive heterobifunctional crosslinking reagent.

Solubility
Soluble in methanol, dimethyl formamide, alcohol and dimethyl sulfoxide. Slightly soluble in water.

Notes
Moisture sensitive. Store in cool place.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7423-55-4
Molecular Formula C7H7NO4
Molecular Weight (g/mol) 169.14
MDL Number MFCD00043030
InChI Key IUTPJBLLJJNPAJ-UHFFFAOYSA-N
Synonym 3-maleimidopropionic acid, 3-2,5-dioxo-2,5-dihydro-1h-pyrrol-1-yl propanoic acid, n-maleoyl-beta-alanine, 3-2,5-dioxopyrrol-1-yl propanoic acid, 1h-pyrrole-1-propanoic acid, 2,5-dihydro-2,5-dioxo, maleimide-ch2 2-cooh, n-carboxyethylmaleimide, 3-maleimdepropionicacid, pubchem11845, acmc-209otw
PubChem CID 573621
IUPAC Name 3-(2,5-dioxopyrrol-1-yl)propanoic acid
SMILES OC(=O)CCN1C(=O)C=CC1=O

Specifications

Melting Point 108°C to 112°C
Color White
Quantity 1 g
Beilstein 1528952
Sensitivity Moisture sensitive
Solubility Information Soluble in methanol,dimethyl formamide,alcohol and dimethyl sulfoxide. Slightly soluble in water.
Formula Weight 169.13
Percent Purity 95%
Physical Form Solid
Chemical Name or Material 3-Maleimidopropionic acid

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'