missing translation for 'onlineSavingsMsg'
Learn More

3-Keto Valproic Acid Sodium Salt, TRC

Product Code. 30397404
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30397404 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30397404 Supplier Toronto Research Chemicals Supplier No. TRCK315500100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 3-Keto Valproic Acid Sodium Salt
CAS 1184991-15-8
Molecular Formula C8 H13 O3 . Na
Quantity 100 mg
InChI Formula InChI=1S/C8H14O3.Na/c1-3-5-6(8(10)11)7(9)4-2;/h6H,3-5H2,1-2H3,(H,10,11);/q;+1/p-1
SMILES [Na+].CCCC(C(=O)[O-])C(=O)CC
IUPAC Name sodium;3-oxo-2-propylpentanoate
Molecular Weight (g/mol) 180.18
Formula Weight 180.0762
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.