missing translation for 'onlineSavingsMsg'
Learn More

3-Keto Fluvastatin Sodium Salt, TRC

Product Code. 30401085
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30401085 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30401085 Supplier Toronto Research Chemicals Supplier No. TRCK18890010MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 3-keto Fluvastatin sodium salt
CAS 1331643-17-4
Molecular Formula C24 H23 F N O4 . Na
Quantity 10 mg
Synonym 6-Heptenoic acid, 7-[3-(4-fluorophenyl)-1-(1-methylethyl)-1H-indol-2-yl]-5-hydroxy-3-oxo-, sodium salt (1:1), Sodium 7-[3-(4-Fluorophenyl)-1-(methylethyl)-1H-indol-2-yl]-5-hydroxy-3-oxo-hept-6-enoate
InChI Formula InChI=1S/C24H24FNO4.Na/c1-15(2)26-21-6-4-3-5-20(21)24(16-7-9-17(25)10-8-16)22(26)12-11-18(27)13-19(28)14-23(29)30;/h3-12,15,18,27H,13-14H2,1-2H3,(H,29,30);/q;+1/p-1
SMILES [Na+].CC(C)n1c(C=CC(O)CC(=O)CC(=O)[O-])c(c2ccc(F)cc2)c3ccccc13
IUPAC Name sodium;7-[3-(4-fluorophenyl)-1-propan-2-ylindol-2-yl]-5-hydroxy-3-oxohept-6-enoate
Molecular Weight (g/mol) 431.43
Formula Weight 431.1509
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.