missing translation for 'onlineSavingsMsg'
Learn More

3-Isopropylphenyl Methylcarbamate, TRC

Product Code. 30401164
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30401164 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30401164 Supplier Toronto Research Chemicals Supplier No. TRCI87494550MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

CAS 64-00-6
Molecular Formula C11H15NO2
Quantity 50 mg
Synonym 3-Isopropylphenyl N-Methylcarbamate, AC 5727, Compound 10854, ENT 25500, H 5727, H 8757, Hercules 5727, Hercules AC 5727, OMS 15, OMS 162, UC 10854, m-Cumenol Methylcarbamate, m-Cumenyl Methylcarbamate, m-Isopropylphenol Methylcarbamate, m-Isopropylphenyl N-Methylcarbamate, m-Isopropylphenyl Methylcarbamate
InChI Formula InChI=1S/C11H15NO2/c1-8(2)9-5-4-6-10(7-9)14-11(13)12-3/h4-8H,1-3H3,(H,12,13)
SMILES CNC(=O)Oc1cccc(c1)C(C)C
IUPAC Name (3-propan-2-ylphenyl) N-methylcarbamate
Molecular Weight (g/mol) 193.24
Formula Weight 193.1103
Chemical Name or Material 3-Isopropylphenyl Methylcarbamate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.