missing translation for 'onlineSavingsMsg'
Learn More

3-Isopropylphenyl Diphenyl Phosphate, TRC

Product Code. 30398607
Change view
Click to view available options
Quantity:
100 mg
1 g
Unit Size:
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30398607 1 g 1g
30402296 100 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30398607 Supplier Toronto Research Chemicals Supplier No. TRCI8729351G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 69515-46-4
Molecular Formula C21 H21 O4 P
Molecular Weight (g/mol) 368.36
Synonym Phosphoric acid, 3-(1-methylethyl)phenyl diphenyl ester (9CI, ACI), 3-(1-Methylethyl)phenyl diphenyl phosphate (ACI), Diphenyl 3-isopropylphenyl phosphate, m-Isopropylphenyl diphenyl phosphate, 3-(1-Methylethyl)phenyl Diphenyl Phosphoric Acid Ester
IUPAC Name (3-isopropylphenyl) diphenyl phosphate
SMILES O=P(OC=1C=CC=CC1)(OC=2C=CC=CC2)OC3=CC=CC(=C3)C(C)C

Specifications

Quantity 1 g
InChI Formula InChI=1S/C21H21O4P/c1-17(2)18-10-9-15-21(16-18)25-26(22,23-19-11-5-3-6-12-19)24-20-13-7-4-8-14-20/h3-17H,1-2H3
Formula Weight 368.1177
Percent Purity >95%
Chemical Name or Material 3-Isopropylphenyl Diphenyl Phosphate
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.