missing translation for 'onlineSavingsMsg'
Learn More

3-Isopropylcatechol, TRC

Product Code. 30396342
Change view
Click to view available options
Quantity:
10 mg
25 mg
100 mg
Unit Size:
100mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30396342 100 mg 100mg
30398627 25 mg -
30390515 10 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30396342 Supplier Toronto Research Chemicals Supplier No. TRCI821995100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2138-48-9
Molecular Formula C9H12O2
Molecular Weight (g/mol) 152.19
Synonym 3-(1-Methylethyl)-1,2-benzenediol, 3-Isopropylpyrocatechol, 1,2-Dihydroxy-3-isopropylbenzene, 3-Isopropyl-1,2-dihydroxybenzene, 3-Isopropylbenzene-1,2-diol, NSC 62675
SMILES CC(C)c1cccc(O)c1O

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C9H12O2/c1-6(2)7-4-3-5-8(10)9(7)11/h3-6,10-11H,1-2H3
Formula Weight 152.0837
Chemical Name or Material 3-Isopropylcatechol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.