missing translation for 'onlineSavingsMsg'
Learn More

3-Isopropylamino-1,2-propanediol, TRC

Product Code. 30390291
Change view
Click to view available options
Quantity:
1 g
10 g
50 g
Unit Size:
10g
1g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30390291 1 g 1g
30405207 10 g 10g
30403053 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30390291 Supplier Toronto Research Chemicals Supplier No. TRCI8240551G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6452-57-9
Molecular Formula C6 H15 N O2
Molecular Weight (g/mol) 133.19
Synonym (2RS)-3-[(1-Methylethyl)amino]propane-1,2-diol, Metoprolol Succinate Imp. N (EP), Metoprolol Tartrate Imp. N (EP)
IUPAC Name 3-(propan-2-ylamino)propane-1,2-diol
SMILES CC(C)NCC(O)CO

Specifications

Quantity 1 g
InChI Formula InChI=1S/C6H15NO2/c1-5(2)7-3-6(9)4-8/h5-9H,3-4H2,1-2H3
Formula Weight 133.1103
Chemical Name or Material (2RS)-3-[(1-Methylethyl)amino]propane-1,2-diol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.