missing translation for 'onlineSavingsMsg'
Learn More

3-Isopropoxybenzeneboronic acid, 97%, Thermo Scientific™

Catalog Number 15489747
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
216485-86-8
C9H13BO3
180.01
MFCD03701512
UKZVUHVNTYDSOP-UHFFFAOYSA-N
3-isopropoxyphenylboronic acid, 3-isopropoxyphenyl boronic acid, 3-isopropoxybenzeneboronic acid, 3-propan-2-yloxyphenyl boronic acid, 3-propan-2-yloxy phenyl boronic acid, 3-1-methylethoxy phenylboronic acid, 3-1-methylethyl oxyphenylboronic acid, 3-iso-propoxyphenylboronic acid, acmc-1ckjx, 5-isopropoxyphenylboronic acid
5083932
(3-propan-2-yloxyphenyl)boronic acid
CC(C)OC1=CC=CC(=C1)B(O)O
This item is not returnable. View return policy
216485-86-8
C9H13BO3
180.01
MFCD03701512
UKZVUHVNTYDSOP-UHFFFAOYSA-N
3-isopropoxyphenylboronic acid, 3-isopropoxyphenyl boronic acid, 3-isopropoxybenzeneboronic acid, 3-propan-2-yloxyphenyl boronic acid, 3-propan-2-yloxy phenyl boronic acid, 3-1-methylethoxy phenylboronic acid, 3-1-methylethyl oxyphenylboronic acid, 3-iso-propoxyphenylboronic acid, acmc-1ckjx, 5-isopropoxyphenylboronic acid
5083932
(3-propan-2-yloxyphenyl)boronic acid
CC(C)OC1=CC=CC(=C1)B(O)O

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Melting Point 76°C to 81°C
Quantity 1 g
Formula Weight 180.01
Percent Purity 97%
Chemical Name or Material 3-Isopropoxybenzeneboronic acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'