missing translation for 'onlineSavingsMsg'
Learn More

3-Iodothyronamine (4-Hydroxyphenyl-13C6) Hydrochloride, TRC

Product Code. 30393751
Change view
Click to view available options
Quantity:
1 mg
2.5 mg
5 mg
Unit Size:
2.5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30393751 1 mg -
30396038 5 mg -
30393750 2.5 mg 2.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30393751 Supplier Toronto Research Chemicals Supplier No. TRCI7205011MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

Molecular Formula C813C6H14INO2 • (HCl)
Molecular Weight (g/mol) 361.133646
Synonym 4-[4-(2-Aminoethyl)-2-iodophenoxy]phenol-13C6 Hydrochloride
SMILES NCCC1=CC=C(O[13C]2=[13CH][13CH]=[13C](O)[13CH]=[13CH]2)C(I)=C1.Cl

Specifications

Quantity 1 mg
InChI Formula InChI=1S/C14H14INO2.ClH/c15-13-9-10(7-8-16)1-6-14(13)18-12-4-2-11(17)3-5-12;/h1-6,9,17H,7-8,16H2;1H/i2+1,3+1,4+1,5+1,11+1,12+1;
Chemical Name or Material 3-Iodothyronamine (4-Hydroxyphenyl-13C6) Hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.