missing translation for 'onlineSavingsMsg'
Learn More

3-Iodothyroacetic Acid, TRC

Product Code. 30399052
Change view
Click to view available options
Quantity:
10 mg
100 mg
Unit Size:
10mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30399052 100 mg -
30402932 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30399052 Supplier Toronto Research Chemicals Supplier No. TRCI728600100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 60578-17-8
Molecular Formula C14 H11 I O4
Molecular Weight (g/mol) 370.139
Synonym 4-(4-Hydroxyphenoxy)-3-iodobenzeneacetic Acid, [4-(p-Hydroxyphenoxy)-3-iodophenyl]acetic Acid
IUPAC Name 2-[4-(4-hydroxyphenoxy)-3-iodophenyl]acetic acid
SMILES OC(=O)Cc1ccc(Oc2ccc(O)cc2)c(I)c1

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C14H11IO4/c15-12-7-9(8-14(17)18)1-6-13(12)19-11-4-2-10(16)3-5-11/h1-7,16H,8H2,(H,17,18)
Formula Weight 369.97
Chemical Name or Material 3-Iodothyroacetic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.