missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Iodopyrazolo[1,5-a]pyridine, 97%
Click to view available options
Quantity:
1 g
Unit Size:
1g
Description
Mostly used in the suzuki coupling reaction.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Mostly used in the suzuki coupling reaction.
Notes
Light Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Mostly used in the suzuki coupling reaction.
Notes
Light Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Specifications
Specifications
| CAS | 19363-99-6 |
| Melting Point | 67°C to 69°C |
| Color | Brown |
| Molecular Formula | C7H5IN2 |
| MDL Number | MFCD20483547 |
| Quantity | 1 g |
| Sensitivity | Light sensitive |
| Synonym | 3-iodopyrazolo 1,5-a pyridine, 3-iodo-pyrazolo 1,5-a pyridine |
| InChI Key | PNNARKUCAKULAL-UHFFFAOYSA-N |
| SMILES | C1=CC2=C(C=NN2C=C1)I |
| Show More |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction