missing translation for 'onlineSavingsMsg'
Learn More

3-Iodopropylene-1-naphthalene Methyl Amine, TRC

Product Code. 30397108
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30397108 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30397108 Supplier Toronto Research Chemicals Supplier No. TRCI71947525G
 Restricted Item
Estimated Shipment: 14-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 3-Iodopropylene-1-naphthalene Methyl Amine
CAS 1076198-32-7
Molecular Formula C14H14IN
Quantity 25 g
Synonym N-(3-Iodo-2-propen-1-yl)-1-naphthalenemethanamine
InChI Formula InChI=1S/C14H14IN/c1-16(11-5-10-15)14-9-4-7-12-6-2-3-8-13(12)14/h2-9,11H,10H2,1H3/b11-5+
SMILES CN(\C=C\CI)c1cccc2ccccc12
IUPAC Name N-[(E)-3-iodoprop-1-enyl]-N-methylnaphthalen-1-amine
Molecular Weight (g/mol) 323.17
Formula Weight 323.0171
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.