missing translation for 'onlineSavingsMsg'
Learn More

3-Iodophthalic acid, ≥95%, Thermo Scientific™

Product Code. 10728625
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10728625 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10728625 Supplier Thermo Scientific Maybridge Supplier No. BTB12976EA
 Restricted Item
Estimated Shipment: 08-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 6937-34-4
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Molecular Formula C8H5IO4
Linear Formula IC6H3(CO2H)2
MDL Number MFCD00275568
Quantity 10 g
Synonym 3-iodobenzene-1,2-dicarboxylic acid, 1,2-benzenedicarboxylicacid, 3-iodo, maybridge1_008994, 3-iodo-phthalic acid, 1,2-benzenedicarboxylic acid, 3-iodo
InChI Key HNPVERUJGFNNRV-UHFFFAOYSA-N
SMILES OC(=O)C1=CC=CC(I)=C1C(O)=O
IUPAC Name 3-iodophthalic acid
Molecular Weight (g/mol) 292.03
PubChem CID 238231
Formula Weight 292.02
Percent Purity 97%
Chemical Name or Material 3-Iodophthalic acid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.