Learn More
Thermo Scientific Maybridge 3-Iodophenylboronic acid, 1g, 97%
Brand: Thermo Scientific Maybridge AC35947DA
3-Iodophenylboronic acid, 1g, 97%
Code : 31
Additional Details : CAS Number : 221037-98-5 Weight : 0.05100kg
Brand: Thermo Scientific Maybridge AC35947DA
3-Iodophenylboronic acid, 1g, 97%
Code : 31
Additional Details : CAS Number : 221037-98-5 Weight : 0.05100kg