missing translation for 'onlineSavingsMsg'
Learn More

3-Iodophenylacetonitrile, 97%

Product Code. 11388146
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11388146 5 g 5g
11378146 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11388146 Supplier Thermo Scientific Alfa Aesar Supplier No. L15118.06
 Restricted Item
Estimated Shipment: 07-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 130723-54-5 | C8H6IN | 243.05 g/mol

3-Iodophenylacetonitrile is a important organic intermediate. It is used in agrochemical, pharmaceutical and dyestuff field.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Iodophenylacetonitrile is a important organic intermediate. It is used in agrochemical, pharmaceutical and dyestuff field.

Solubility
Insoluble in water

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. It is sensitive to light. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 130723-54-5
Molecular Formula C8H6IN
Molecular Weight (g/mol) 243.05
MDL Number MFCD00040890
InChI Key LVOKGAHCTHNWIL-UHFFFAOYSA-N
Synonym 3-iodophenylacetonitrile, 2-3-iodophenyl acetonitrile, 3-iodobenzyl cyanide, 3-iodophenyl acetonitrile, m-iodobenzyl cyanide, 3-iodophenyacetonitrile, pubchem12771, 3-iodo-1-phenylacetonitrile, acmc-1c0v0, benzeneacetonitrile, 3-iodo
PubChem CID 2759368
IUPAC Name 2-(3-iodophenyl)acetonitrile
SMILES IC1=CC=CC(CC#N)=C1

Specifications

Melting Point 33°C to 35°C
Boiling Point 138°C (12 mmHg)
Flash Point >110°C (230°F)
Quantity 5 g
UN Number UN3439
Beilstein 7350695
Sensitivity Light sensitive
Solubility Information Insoluble in water
Formula Weight 243.05
Percent Purity 97%
Chemical Name or Material 3-Iodophenylacetonitrile
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.