missing translation for 'onlineSavingsMsg'
Learn More

3-Iodophenyl isocyanate, 97%

Product Code. 15463987
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 15463987

Brand: Thermo Scientific Alfa Aesar H30803.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 23138-56-9 | C7H4INO | 245.019 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 23138-56-9
Molecular Formula C7H4INO
Molecular Weight (g/mol) 245.019
MDL Number MFCD00037088
InChI Key LZSQANBVKPXBLO-UHFFFAOYSA-N
Synonym 3-iodophenyl isocyanate, benzene,1-iodo-3-isocyanato, 3-iodophenylisocyanate, 1-iodo-3-isocyanato-benzene, acmc-20an25
PubChem CID 4499000
IUPAC Name 1-iodo-3-isocyanatobenzene
SMILES C1=CC(=CC(=C1)I)N=C=O

Specifications

Boiling Point 55°C (0.2 mmHg)
Flash Point >110°C (230°F)
Odor Pungent
Quantity 1 g
UN Number UN2206
Formula Weight 245.02
Percent Purity 97%
Chemical Name or Material 3-Iodophenyl isocyanate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.